Auftragserfassung
Switzerland
ContactUsLinkComponent
Triethanolamin (Trolamin, TEA), BAKER ANALYZED®, Laborreagens, J.T.Baker®
Triethanolamin (Trolamin, TEA), BAKER ANALYZED®, Laborreagens, J.T.Baker®
  7078.9025
 :  Avantor
 :  
undefined
Triethanolamin (Trolamin, TEA), BAKER ANALYZED®, Laborreagens, J.T.Baker®
  7078.9025
 :  Avantor
 :  

 

  • Packung:
    Kunststoffkanister
  • VE:
    25 L
  • Seq:
    5327

 

 

  • Assay:
    99.0 - 103.0 %
  • Appearance of solution:
    passes test
  • Density (g/ml) at 25°C:
    1.117 - 1.125
  • Iron (Fe):
    max. 5 ppm
  • Heavy Metals (as Pb):
    max. 10 ppm
  • Identification:
    passes test
  • Sulfated Ash:
    max. 0.005 %
  • Water (H2O):
    max. 0.2 %

 

 

 :
 :149.19 g/mol
 :13,09739
 :335,4 °C (1013 hPa)
 :21,6 °C
 :1,13 g/cm³ (20 °C)
 :179 °C (geschlossener Tiegel)
 :Raumtemperatur
 :203-049-8
 :102-71-6
 :C₆H₁₅NO₃
 :MFCD00002855