


N,N-Dimethylformamide dimethyl acetal for GC derivatisation, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:4637-24-5
:
Formula:C₅H₁₃NO₂
MDL Number:MFCD00008482
:119.16 g/mol
Boiling Pt:102…103 °C (720 mmHg) (lit.)
Density:0.897 g/mL (25 °C) (lit.)
Flash Pt:5 °C
EINECS:225-063-3
ADR:3,II
:M-Clarity™ quality level = MQ100
UN:3271
:GHS02
:GHS07
:GHS08
Specification Test Results
- APPEARANCE:COLORLESS LIQUID
- INFRARED SPECTRUM:CONFORMS TO STRUCTURE
- GAS LIQUID:CA. 95%
Specification Test Results
Frequently Bought Together