



N,N-Dimethylformamide dimethyl acetal for synthesis, Sigma-Aldrich®
: Merck
:4637-24-5
:
:C₅H₁₃NO₂
:MFCD00008482
:119.16 g/mol
:102 °C (1013 hPa)
:0.893 g/cm³ (20 °C)
:6 °C
:Ambient
:225-063-3
:3,II
:M-Clarity™ quality level = MQ200
:1993
:GHS02
:GHS05
:GHS08
:GHS07
- Solubility:(20 °C) (slow decomposition)
- Melting Point:-85 °C
- Molar Mass:119.16 g/mol
- Boiling Point:107 - 108 °C
- Flash Point:6 °C
- Refractive Index:1.3972 (20 °C, 589 nm)
- Explosion Limit:1.3 - 17.7 %(V)
- Density:0.90 g/cm3 (20 °C)
- pH:7 (H2O)
- Ignition Point:155 °C
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Frequently Bought Together