VWR®, Hoods with Integrated Face Mask
Supplier: VWR Collection
Certificates
About this item
These disposable hoods, with 2 ply face mask, are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Free from natural rubber latex
- Available in a wide range of colours
Weight: 10 g/m²
Packaging: 50 pieces per bag, 10 bags per case.
Specifications
- Catalog No:
- 113-8911
- 113-8912
- Garment application:
- Laboratory
- Laboratory
- Weight:
- 10 g/m²
- 10 g/m²
Specifications
Related Information
Food contact certificate F
Additional Product Details
TDS VWR® Hoodcap with Face Mask
Datasheet
VWR® Non Regulated Disposables for Production
Product Literature
Frequently Bought Together