
Triethylsilyl trifluoromethanesulphonate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79271-56-0
:
Formula:C₇H₁₅F₃O₃SSi
MDL Number:MFCD00000407
Boiling Pt:207 °C
Density:1.225
Flash Pt:72 °C (161 °F)
EINECS:279-124-4
UN:3265
ADR:8,II
:264.34 g/mol
:GHS05
Related Information
Frequently Bought Together