Order Entry
Switzerland
ContactUsLinkComponent
1,1,1,3,3,3-Hexafluoro-2-propanol 99.8% for peptide synthesis
Corrosive
Irritant
1,1,1,3,3,3-Hexafluoro-2-propanol 99.8% for peptide synthesis
Catalog # ACRO354861000
CAS Number:  
undefined
Corrosive
Irritant
1,1,1,3,3,3-Hexafluoro-2-propanol 99.8% for peptide synthesis
Catalog # ACRO354861000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.02 % (K.F.)
  • GC:
    ≥99.8 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0005 %
  • Free amine:
    ≤0.0002 %

Specification Test Results

About this item

 :
Merck Index:13,09598
Boiling Pt:59.1 °C (1013 hPa)
Melting Pt:–4 °C
Density:1.596 g/cm³
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:213-059-4
UN:3265
ADR:8,I
 :168.04 g/mol
 :GHS05
 :GHS07
CAS Number:920-66-1
Formula:C₃H₂F₆O
MDL Number:MFCD00011651