
EDTA (ethylenediamine tetraacetic acid), Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:60-00-4
:
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541
:292.25 g/mol
Merck Index:13,03546
Boiling Pt:614 °C (1013 hPa)
Melting Pt:220 °C
Density:0.86 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:200-449-4
REACH:01-2119486399-18
:GHS07
Specification Test Results
- Solubility:0.5 g/l (20 °C)
- Melting Point:220 °C (decomposition)
- Molar Mass:292.24 g/mol
- Bulk Density:600 kg/m3
- Vapor Pressure:- 0.013 hPa (20 °C)
- Flash Point:100 °C DIN 51758
- Density:0.86 g/cm3 (20 °C)
- pH:2.5 (10 g/l, H2O, 23 °C) (slurry)
- Ignition Point:200 °C
Specification Test Results
Frequently Bought Together