Order Entry
Switzerland
ContactUsLinkComponent
 
Potassium citrate monohydrate ≥99%, GPR RECTAPUR®
 
undefined
Potassium citrate monohydrate ≥99%, GPR RECTAPUR®
 

 

  • Assay:
    Min. 99 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Cl (Chloride):
    50 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • Fe (Iron):
    Max. 10 ppm

 

Frequently Bought Together