Order Entry
Switzerland
ContactUsLinkComponent
di-Potassium oxalate monohydrate 99.5-101.0%, AnalaR NORMAPUR® analytical reagent
Irritant
di-Potassium oxalate monohydrate 99.5-101.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 26887.293
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
di-Potassium oxalate monohydrate 99.5-101.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 26887.293
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    99.5 - 101.0 %
  • pH (20°C; 5 %):
    7.0 - 8.5
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Insolubility in water:
    Max. 100 ppm
  • Cl (Chloride):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Na (Sodium):
    Max. 0.02 %

Specification Test Results

About this item

 :
 :184.23 g/mol
Merck Index:12,07820
Melting Pt:356 °C
Density:2.154 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-506-8
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:6487-48-5
Formula:(COOK)₂·H₂O
MDL Number:MFCD00150033