
di-Potassium oxalate monohydrate 98.5-101.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6487-48-5
:
Formula:(COOK)₂·H₂O
MDL Number:MFCD00150033
:184.23 g/mol
Merck Index:14,7651
Melting Pt:>300 °C
Density:2.13
Storage Temperature:Ambient
EINECS:209-506-8
UN:2811
ADR:6.1,III
:GHS07
Related Information
Frequently Bought Together