Chemical resistant suit, Microchem® by AlphaTec® 5000, model 151-G02
Supplier: Ansell
Certificates
About this item
Highly visible rear entry suit, with neoprene rubber face seal for a close fit to full face respirators. Ideal for use in extremly hazardous areas where protection against concentrated chemicals and biological agents is required.
- Neoprene rubber face seal
- Rear horizontal zip entry
- Attached socks with boot overflap
- Attached Ansell Barrier gloves, with oversleeves and finger loops
- Ultrasonically welded and taped seams provide a high barrier to liquid and particulates
- Bright orange colour for improved worker safety
- Multi-layer material is lightweight, yet strong and durable
Typical applications include: Industrial and tank cleaning, sewage purification installations, chemicals, oil and petrochemicals, pharmaceuticals, agriculture, HAZMAT Emergency Response (i.e. Level B), first response and fire service.
Certifications: Type 3/4/5, EN 14126, EN 1073-2, EN 1149-5; Cat. III
Specifications
- Catalog No:
- ANSEOR50T-15106G02
- ANSEOR50T-15105G02
- ANSEOR50T-15107G02
- ANSEOR50T-15102G02
- ANSEOR50T-15104G02
- ANSEOR50T-15103G02
- ANSEOR50T-15108G02
- Model:
- 5000, 151-G02
- 5000, 151-G02
- 5000, 151-G02
- 5000, 151-G02
- 5000, 151-G02
- 5000, 151-G02
- 5000, 151-G02
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial