Order Entry
Switzerland
ContactUsLinkComponent
di-Sodium succinate hexahydrate for synthesis, Sigma-Aldrich®
di-Sodium succinate hexahydrate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
di-Sodium succinate hexahydrate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    349 g/l (25 °C)
  • Melting Point:
    120 °C Elimination of water of crystallisation
  • Molar Mass:
    270.15 g/mol
  • pH:
    6.0 - 8.5 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together