Order Entry
Switzerland
ContactUsLinkComponent
 
1,16-Hexadecanedioic acid for synthesis, Sigma-Aldrich®
  8.41740.0001
 :  Merck
 :  
1,16-Hexadecanedioic acid for synthesis, Sigma-Aldrich®
  8.41740.0001
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Melting Point:
    122 - 123 °C
  • Molar Mass:
    286.41 g/mol
  • Hill Formula:
    C16H30O4

 

 

 :
 :430 °C (1013 hPa)
 :122…123 °C
 :M-Clarity™ quality level = MQ200
 :286.41 g/mol
 :Ambient
 :208-013-5
 :505-54-4
 :C₁₆H₃₀O₄
 :MFCD00002746