Order Entry
Switzerland
ContactUsLinkComponent
L-Methionine for synthesis, Sigma-Aldrich®
L-Methionine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
L-Methionine for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    48 g/l (20 °C)
  • Melting Point:
    280 - 285 °C
  • Molar Mass:
    149.2 g/mol
  • Bulk Density:
    550 kg/m3
  • pH:
    5 - 7 (10 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together