Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Glycine

Glycine

2.000 Assays
Catalog # CHMCAB5020
Supplier:   Chemicon
Glycine
Catalog # CHMCAB5020
Supplier:  Chemicon
CAS Number:  56-40-6
Glycine
Glycine

2.000 Assays

CAS Number:  56-40-6
Price
1 460.00 SFr.
Log in for availability
1 * 2.000 Essai
Product Details & Documents
Synonyms:  Aminoacetic acid, Glycocoll, H-Gly-OH
  • CAS Number:56-40-6
  • Formula:NH₂CH₂COOH
  • MDL Number:MFCD00008131
  • Molecular Weight:75.07 g/mol
  • Merck Index:13,04504
  • Boiling Pt:181 °C (1013 hPa)
  • Melting Pt:83 °C
  • Density:1.64 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-272-2
Specifications

Specifications

  • Pk:
    2.000 Assays

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.