
6-Aminocaproic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:60-32-2
:
Formula:C₆H₁₃NO₂
MDL Number:MFCD00008238
:131.17 g/mol
Merck Index:13,00431
Boiling Pt:257 °C (1013 hPa)
Melting Pt:127 °C
:M-Clarity™ quality level = MQ200
Flash Pt:207 °C
EINECS:200-469-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:800 g/l (20 °C)
- Molar Mass:131.17 g/mol
- Flash Point:207 - 209 °C
- pH:7.0 - 7.5 (50 g/l, H2O, 20 °C)
Specification Test Results
Frequently Bought Together