4-Aminobutyric acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:56-12-2
:
Formula:C₄H₉NO₂
MDL Number:MFCD00008226
:103.12 g/mol
Merck Index:13,00429
Boiling Pt:248 °C (1013 hPa)
Melting Pt:202 °C
Density:1.21 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-258-6
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:200 - 202 °C
- Molar Mass:103.12 g/mol
Specification Test Results
Related Information
msds_800302 msds_800302
Sicherheitsdatenblatt (SDS)
ProductPage_800302 ProductPage_800302
Produktspezifikation
Frequently Bought Together