


Mercury(II) acetate ACS, Reag. Ph. Eur. analytical reagent, Supelco®
: Merck
:1600-27-7
:
:(H₃CCOO)₂Hg
:MFCD00012165
:318.68 g/mol
:13,05898
:178…180 °C
:3.27 g/cm³ (20 °C)
:Ambient
:216-491-1
:6.1,II
:M-Clarity™ quality level = MQ300
:1629
:GHS06
:GHS08
:GHS09
- Solubility:400 g/l (20 °C)
- Melting Point:178 - 180 °C (decomposition)
- Molar Mass:318.68 g/mol
- Bulk Density:1000 kg/m3
- Density:3.27 g/cm3 (20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Frequently Bought Together