Order Entry
Switzerland
ContactUsLinkComponent
N-Lauroylsarcosine sodium salt for synthesis, Sigma-Aldrich®
Toxic
Corrosive
N-Lauroylsarcosine sodium salt for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Toxic
Corrosive
N-Lauroylsarcosine sodium salt for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    45 - 47 °C
  • Molar Mass:
    293.38 g/mol
  • Bulk Density:
    400 kg/m3
  • Flash Point:
    150 °C
  • pH:
    8 (30 g/l, H2O, 20 °C)
  • Synonyms:
    Sarcosyl, Sodium lauroylsarcosinate
  • Hill Formula:
    C15H28NNaO3
  • Chemical Formula:
    CH3-(CH2)10-CO-N(CH3)-CH2-COONa

Specification Test Results

Related Information

Frequently Bought Together