Order Entry
Switzerland
ContactUsLinkComponent
Sodium acetate ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Sodium acetate ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Sodium acetate ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Identification:
    Passes test
  • IR Spectrum:
    Passes test
  • pH (25°C; 5 %):
    7.0 - 9.2
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Insolubility in water:
    Max. 100 ppm
  • Loss on drying (120°C):
    Max. 1.0 %
  • Cl (Chloride):
    Max. 20 ppm
  • PO4 (Phosphate):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 30 ppm
  • Al (Aluminium):
    Max. 10 ppm
  • Ca (Calcium):
    Max. 50 ppm
  • Cu (Copper):
    Max. 3 ppm
  • Fe (Iron):
    Max. 10 ppm
  • K (Potassium):
    Max. 0.05 %
  • Mg (Magnesium):
    Max. 20 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together