Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cesium acetate
Cesium acetate ≥99% (metals basis)
Cesium acetate ≥99% (metals basis)
:
Thermo Fisher Scientific
Cesium acetate ≥99% (metals basis)
:
Thermo Fisher Scientific
:
3396-11-0
:
:
H₃CCOOCs
:
MFCD00013056
:
191.95 g/mol
:
194 °C
:
Ambient
:
222-248-0
msds_ALFA043095
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone, EMPARTA® ACS analytical reagent, Supelco®
: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
: VWR Chemicals
2,2,4-Trimethylpentane, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Ethanol absolute ≥99.9% (by GC), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
2-Propanol ≥99.9% (by GC), LiChrosolv®, hypergrade for LC-MS, Supelco®
: Merck