
Cesium acetate 99%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3396-11-0
:
Formula:H₃CCOOCs
MDL Number:MFCD00013056
:191.95 g/mol
Melting Pt:194 °C
EINECS:222-248-0
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder and/or chunks
- Titration with HClO4:≥98.5 %
Specification Test Results
Frequently Bought Together