
Sodium butyrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:156-54-7
:
Formula:C₄H₇NaO₂
MDL Number:MFCD00002816
:110.09 g/mol
Melting Pt:>300 °C
Density:1.324 g/cm³ (30 °C)
EINECS:205-857-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together