Order Entry
Switzerland
ContactUsLinkComponent
Acetic acid glacial 99.5-100.05% USP, Multi-Compendial, J.T.Baker®
Acetic acid glacial 99.5-100.05% USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Acetic acid glacial 99.5-100.05% USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    = 15.6 °C
  • USP - Identification:
    Passes Test
  • USP - Congealing Temperature:
    ≥15.6 °C
  • USP - Nonvolatile Residue (USP), mg/20 mL:
    ≤1.0
  • USP - Chloride (Cl):
    Passes Test
  • USP - Sulfate (SO₄):
    Passes Test
  • USP - Readily Oxidizable Substances:
    Passes Test
  • USP - Assay (CH₃COOH):
    99.5 - 100.5 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance:
    Passes Test
  • EP/BP - Freezing Point:
    ≥14.8 °C
  • EP/BP - Iron (Fe):
    ≤5 ppm
  • EP/BP - Chloride (Cl):
    ≤25 ppm
  • EP/BP - Sulfate (SO₄):
    ≤50 ppm
  • EP/BP - Reducing Substances:
    Passes Test
  • EP/BP - Residue on Evaporation:
    ≤0.01 %
  • EP/BP - Assay (CH₃COOH):
    99.0 - 100.5 %
  • JP - Assay (CH₃COOH):
    99.0 - 101.0 %
  • JP - Identification:
    Passes Test
  • JP - Congealing Point:
    ≥14.5 °C
  • JP - Nonvolatile Residue, mg/10 mL:
    ≤1.0
  • JP - Chloride (Cl):
    Passes Test
  • JP - Sulfate (SO₄):
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤10 ppm
  • JP - Reducing Substances:
    Passes Test

Specification Test Results

Frequently Bought Together