ContactUsLinkComponent
Red 650 dUTP
Red 650 dUTP
Catalog # ENZOENZ42522
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Red 650 dUTP
Catalog # ENZOENZ42522
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    25 Nmo
  • Description:
    Red 650 dUTP, 1 mM, 25 nmol
  • Seq:
    02
Specifications
About this item
Red 650 [Cyanine-5E] dUTP is a cyanine-5 derivative with enhanced spectral properties. This nucleotide analog can replace TTP in reactions in which it serves as a substrate for E.coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase. Fluorescently labeled probes can be prepared with this fluorescent nucleotide by a variety of methods including nick translation, cDNA labelling and 3'-end labelling.
Probes generated by these methods are suitable for use for the identification of specific sequences by in situ hybridisation procedures on fixed cells and tissues by direct fluorescence detection. Red 650 dUTP can also be used for multicolour fluorescence labelling and with the Nick translation DNA labelling system 2,0.