Ethyl acetate
Glass bottle, 2,5 L
Catalog # ROTH7336.1
Marketsource
Supplier: Roth Carl
Ethyl acetate
Catalog # ROTH7336.1
Supplier: Roth Carl
Marketsource
CAS Number: 141-78-6
Jump to:
Product Details & Documents
Synonyms: Acetic acid ethyl ester
- CAS Number:141-78-6
- Formula:CH₃COOC₂H₅
- MDL Number:MFCD00009171
- Molecular Weight:88.11 g/mol
- Merck Index:13,03792
- Boiling Pt:77.1 °C (1013 hPa)
- Melting Pt:–83 °C
- Density:0.902 g/cm³ (20 °C)
- Flash Pt:–4 °C
- Storage Temperature:Ambient
- EINECS:205-500-4
- UN:1173
- ADR:3,II
- REACH:01-2119475103-46
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Cat. No.:ROTH7336.1
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health



