Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Acetanilide ≥98%

Acetanilide ≥98%

Acetanilide ≥98%
Certificates
Acetanilide ≥98%
Acetanilide ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 103-84-4 Molecular Formula: C8H9NO Formula Weight: 135.17 Flash Point: 161°(321°F) Physical Form: Crystals Density: 1.121 Melting Point: 113-116° Boiling Point: 304-305° Beilstein Reference: 606468 Merck Reference: 14,50 MDL No.: MFCD00008674

Synonyms:  N-Phenylacetamide
  • CAS Number:103-84-4
  • Formula:C₈H₉NO
  • MDL Number:MFCD00008674
  • Molecular Weight:135.17 g/mol
  • Merck Index:14,50
  • Boiling Pt:304…305 °C
  • Melting Pt:113…116 °C
  • Density:1.121
  • Flash Pt:161 °C (321 °F)
  • Storage Temperature:Ambient
  • EINECS:203-150-7
Product Family Options

Product Information