N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES) ≥99%, high purity
Catalog # 0285-100G
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Abs.@280 nm (5%, Water):≤0.02
- pH (1%, Water) @25 °C:3.6 – 4.4
- pKa @25 °C:6.58 – 6.98
- Purity:≥99%
- Water (Karl Fisher):≤1%
Specification Test Results
About this item
:
:182.2 g/mol
Merck Index:13,00037
Melting Pt:270…295 °C
Storage Temperature:Ambient
EINECS:230-908-4
CAS Number:7365-82-4
Formula:H₂NCOCH₂NHCH₂CH₂SO₃H
MDL Number:MFCD00008030