N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES) 99%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7365-82-4
:
Formula:H₂NCOCH₂NHCH₂CH₂SO₃H
MDL Number:MFCD00008030
:182.2 g/mol
Merck Index:13,00037
Melting Pt:270…295 °C
Storage Temperature:Ambient
EINECS:230-908-4
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤1 % (K.F.)
- UV:(5 % in water)
- pH:3.6 to 4.4 (1 % solution)
- Titration with NaOH:≥98.5 %
Specification Test Results
Frequently Bought Together