Order Entry
Switzerland
ContactUsLinkComponent
Formamide, (max. 0.1% H₂O) ≥99%, GPR RECTAPUR® for synthesis
Health
Formamide, (max. 0.1% H₂O) ≥99%, GPR RECTAPUR® for synthesis
Catalog # 24312.294
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Formamide, (max. 0.1% H₂O) ≥99%, GPR RECTAPUR® for synthesis
Catalog # 24312.294
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Ignition residue (SO4):
    Max. 100 ppm
  • Water:
    Max. 0.1 %

Specification Test Results

About this item

 :
Merck Index:13,04261
Boiling Pt:210 °C (1013 hPa)
Melting Pt:2 °C
Density:1.1334 g/cm³ (20 °C)
Flash Pt:79 °C
 :45.04 g/mol
Storage Temperature:Refrigerator
EINECS:200-842-0
 :GHS08
CAS Number:75-12-7
Formula:HCONH₂
MDL Number:MFCD00007941