Formamide
Glass bottle, 200 mL
Catalog # NADIEC-608-200ML
Marketsource
Supplier: NATIONAL DIAGNOSTICS
Jump to:
Product Details & Documents
Synonyms: Carbamaldehyde
- CAS Number:75-12-7
- Formula:HCONH₂
- MDL Number:MFCD00007941
- Molecular Weight:45.04 g/mol
- Merck Index:13,04261
- Boiling Pt:210 °C (1013 hPa)
- Melting Pt:2 °C
- Density:1.1334 g/cm³ (20 °C)
- Flash Pt:79 °C
- Storage Temperature:Refrigerator
- EINECS:200-842-0
Specifications
Specifications
- Pack type:Glass bottle
- Pk:200 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



