
Formamide, EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:75-12-7
:
Formula:HCONH₂
MDL Number:MFCD00007941
Merck Index:13,04261
Boiling Pt:103 °C (2 kPa)
Melting Pt:2 °C
Density:1.1334 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Flash Pt:79 °C
:45.04 g/mol
EINECS:200-842-0
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Assay (N; Kjeldahl):≥ 99.2 %
- Identity (IR):passes test
- Appearance:Clear, colorless liquid.
- Colour (APHA):≤ 10 APHA
- Density (d 20 °C/ 4 °C):1.132 - 1.135
- Solidification point:2.0 - 3.0 °C
- Boiling point (2 kPa):about 103
- Chloride (Cl):≤ 1 ppm
- Cd (Cadmium):≤ 1 ppm
- Cu (Copper):≤ 1 ppm
- Fe (Iron):≤ 1 ppm
- Pb (Lead):≤ 1 ppm
- Zn (Zinc):≤ 1 ppm
- Formic acid:≤ 0.02 %
- Ammonium Formate:≤ 0.1 %
- Sulfated ash (600 °C):≤ 0.005 %
- Water (according to Karl Fischer):≤ 0.1 %
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_109684
SDS
ProductPage_109684 ProductPage_109684
Product specification
Frequently Bought Together