Order Entry
Switzerland
ContactUsLinkComponent
N,N-Dimethylformamide ≥99.5%, GPR RECTAPUR®
Flammable
Health
Irritant
N,N-Dimethylformamide ≥99.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Health
Irritant
N,N-Dimethylformamide ≥99.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Appearance:
    Clear colourless liquid
  • IR Spectrum:
    Passes test
  • Acidity:
    Max. 0.00025 meq/g
  • Alkalinity:
    Max. 0.003 meq/g
  • Boiling point:
    152 - 155 °C
  • Density (20/4):
    0.940 - 0.950
  • Refractive index (20°C):
    1.429 - 1.431
  • Evaporation residue:
    Max. 100 ppm
  • Water:
    Max. 0.1 %

Specification Test Results

Frequently Bought Together