Order Entry
Switzerland
ContactUsLinkComponent
N,N-Dimethylacetamide ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Health
Irritant
N,N-Dimethylacetamide ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Catalog # 83636.350
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Irritant
N,N-Dimethylacetamide ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Catalog # 83636.350
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.50 %
  • Evaporation residue:
    Max. 0.0005 %
  • Water:
    Max. 500 ppm
  • Transmittance (270 nm):
    Min. 10.0 %
  • Transmittance (290 nm):
    Min. 98.0 %
  • Transmittance (310 nm):
    Min. 89.00 %
  • Transmittance (320 nm):
    Min. 94.0 %
  • Transmittance (400 nm):
    Min. 98.0 %

Specification Test Results

About this item

 :
 :87.12 g/mol
Merck Index:13,03254
Boiling Pt:166.1 °C (1013 hPa)
Melting Pt:–20 °C
Density:0.937 g/cm³ (20 °C)
Flash Pt:70 °C
Storage Temperature:Ambient
EINECS:204-826-4
UN:2810
ADR:6.1,III
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
REACH:01-2119459339-27
 :GHS08
 :GHS07
CAS Number:127-19-5
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686