

N,N-Dimethylacetamide 99+% for spectroscopy
:127-19-5
:
:CH₃CON(CH₃)₂
:MFCD00008686
:87.12 g/mol
:13,03254
:164…166 °C (760 mmHg)
:–20 °C
:0.937 g/cm³
:70 °C
:Ambient
:204-826-4
:6.1,III
:GHS07
:GHS08
- Appearance:Clear liquid
- Water:≤0.05 %
- GC:≥99.0 %
- UV:at 270 nm A: ≤1
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
Frequently Bought Together