Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
 
Oxalyl dichloride for synthesis, Sigma-Aldrich®
 :  Merck
 
 
 
 
 
Oxalyl dichloride for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) (rigorous decomposition)
  • Melting Point:
    -12 °C
  • Molar Mass:
    126.92 g/mol
  • Boiling Point:
    63 - 64 °C (1013 hPa)
  • Vapor Pressure:
    200 hPa (20 °C)
  • Flash Point:
    100 °C
  • Density:
    1.48 g/cm3 (20 °C)
  • Hazard Class:
    4.3
  • Hazard Class Description:
    Dangerous When Wet Material

 

Frequently Bought Together