ContactUsLinkComponent
Fluorescein-12-UTP
Fluorescein-12-UTP
Catalog # ENZOENZ42834
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Fluorescein-12-UTP
Catalog # ENZOENZ42834
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    250 Nmo
  • Description:
    Fluorescein-12-UTP, 10 mM, 250 nmol
  • Seq:
    02
Specifications
About this item
Fluorescein-12-UTP is ideal for in vitro transcription reactions catalysed by T3, T7, or SP6 RNA polymerases and reverse transcriptase to produce fluorophore-labeled RNA transcripts. RNA-based probes can be used to identify complementary sequences by in situ hybridisation to fixed cells and tissues by direct or indirect fluorescence detection or by membrane hybridisation procedures.