SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
2-Octanone
2-Octanone ≥98%
2-Octanone ≥98%
:
Thermo Fisher Scientific
2-Octanone ≥98%
:
Thermo Fisher Scientific
:
111-13-7
:
:
C₈H₁₆O
:
MFCD00009540
:
128.21 g/mol
:
14,4711
:
172…173 °C
:
–16 °C
:
0.819
:
54 °C (129 °F)
:
Ambient
:
203-837-1
:
1224
:
3,III
:
GHS02
:
GHS07
msds_ALFAA11643
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Acetone, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Formaldehyde ≥37% in aqueous solution stabilised, GR ACS, Reag. Ph. Eur. analytical reagent, Sigma-Aldrich®
: Merck
Ethanol denatured, EMSURE® for analysis (denatured with 1% MEK), Supelco®
: Merck
(±)-2-Ethyl-1-hexanol for synthesis, Sigma-Aldrich®
: Merck
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2-Hexanone ≥98%
: Thermo Fisher Scientific