Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

2-Chloroethyl isocyanate

Glass bottle for solids, 25 g
Catalog # MOLEM56831722
Marketsource
Supplier:   Molekula
2-Chloroethyl isocyanate
Catalog # MOLEM56831722
Supplier:  Molekula
Marketsource
CAS Number:  1943-83-5
2-Chloroethyl isocyanate

Glass bottle for solids, 25 g

CAS Number:  1943-83-5
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Isocyanic acid 2-Chloroethyl ester
  • CAS Number:1943-83-5
  • Formula:C₃H₄ClNO
  • MDL Number:MFCD00002043
  • Molecular Weight:105.52 g/mol
  • Boiling Pt:135 °C (1013 hPa)
  • Density:1.24 g/cm³ (15 °C)
  • Storage Temperature:Refrigerator
  • EINECS:217-734-4
  • UN:3080
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications