Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Dichloromethane

Dichloromethane

Health
Irritant
Marketsource
Supplier:   Roth Carl
Dichloromethane
Supplier:  Roth Carl
Marketsource
Certificates
Dichloromethane

5 Options Available Below

Product Details & Documents
Synonyms:  Methylene chloride, DCM
  • CAS Number:75-09-2
  • Formula:CH₂Cl₂
  • MDL Number:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck Index:12,06140
  • Boiling Pt:39.8 °C (1013 hPa)
  • Melting Pt:–95 °C
  • Density:1.322 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-838-9
  • UN:1593
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - SFr. 16.20
    SFr. 16.20
    1 * 100 mL
       SFr. 16.20

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - SFr. 35.90
    SFr. 35.90
    1 * 1 L
       SFr. 35.90

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - SFr. 71.40
    SFr. 71.40
    1 * 2,5 L
       SFr. 71.40

    Specifications:
    Marketsource
    Marketsource
  • 10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - SFr. 210.00
    SFr. 210.00
    1 * 10 L
       SFr. 210.00

    Specifications:
    Marketsource
    Marketsource
  • 25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - SFr. 406.80
    SFr. 406.80
    1 * 25 L
       SFr. 406.80

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.