

Dichloromethane ≥99.9% (by GC) stabilised, Suprasolv® hypergrade for organic trace analysis, Supelco®
Catalog # 1.06454.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Purity (GC):≥ 99,9%
- Identity (IR):conforms
- Evaporation Residue:≤3,0 mg/l
- Water:≤0,005%
- Colour:≤10 Hazen
- GC/ECD (Retention Range 1,2,4-Tri- Chlorobenzene to Decachlorobiphenyle, Individual Signals (Lindane Standard)):≤2 pg/ml
- GC/FID (Retention Range n-Undecane to n-Tetracontane, Individual Signals (n-Tetradecane Standard)):≤2 ng/ml
- GC/MSD (Retention Range n-Undecane to n-Tetracontane, Scaning Area 30 - 600 amu, Individual Signals (n-Tetradecane Standard)):≤2 ng/ml
Specification Test Results
About this item
:
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:Stabilised with 2-methyl-2-butene (iso-amylene)
EINECS:200-838-9
UN:1593
:GHS08
:GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881