ContactUsLinkComponent
Pelargonaldehyde analytical standard, Supelco®
Pelargonaldehyde analytical standard, Supelco®
  442719.
 :  Merck
 :  
undefined
Pelargonaldehyde analytical standard, Supelco®
  442719.
 :  Merck
 :  
  • Pk:
    1
  • Seq:
    0001
 :
 :142.24 g/mol
 :190 °C (1013 hPa)
 :–40 °C
 :0.83 g/cm³ (20 °C)
 :76 °C
 :Refrigerator
 :204-688-5
 :124-19-6
 :C₉H₁₈O
 :MFCD00007030