ContactUsLinkComponent
2-Bromoethanol analytical standard, Supelco®
2-Bromoethanol analytical standard, Supelco®
  442334.
 :  Merck
 :  
undefined
2-Bromoethanol analytical standard, Supelco®
  442334.
 :  Merck
 :  
  • Pk:
    1
  • Seq:
    0001
 :
 :124.97 g/mol
 :13,03826
 :149 °C (1013 hPa)
 :–80 °C
 :1.76 g/cm³ (20 °C)
 :40 °C
 :Refrigerator
 :208-748-1
 :2927
 :6.1,II
 :GHS02
 :GHS07
 :540-51-2
 :BrH₂CCH₂OH
 :MFCD00002827