

2-Bromoethanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:540-51-2
:
Formula:BrH₂CCH₂OH
MDL Number:MFCD00002827
:124.97 g/mol
Merck Index:13,03826
Boiling Pt:149 °C (1013 hPa)
Melting Pt:–80 °C
Density:1.76 g/cm³ (20 °C)
EINECS:208-748-1
UN:2929
ADR:6.1 (3),II
:M-Clarity™ quality level = MQ200
Flash Pt:40 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-80 °C
- Molar Mass:124.97 g/mol
- Boiling Point:56 - 57 °C (27 hPa)
- Vapor Pressure:3.2 hPa (20 °C)
- Flash Point:40 °C
- Refractive Index:1.492 (20 °C, 589 nm)
- Density:1.76 g/cm3 (20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
Frequently Bought Together