

1-Bromo-2-methoxyethane 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6482-24-2
:
Formula:BrH₂CCH₂OCH₃
MDL Number:MFCD00000236
:138.99 g/mol
Boiling Pt:107…110 °C (760 mmHg)
Density:1.46 g/cm³
Flash Pt:28 °C
Storage Temperature:Refrigerator
EINECS:229-339-4
UN:1993
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4460 to 1.4500 (20°C, 589 nm)
- GC:≥94 %
Specification Test Results
Frequently Bought Together