Order Entry
Switzerland
ContactUsLinkComponent
L(+)-Histidine
L(+)-Histidine
Catalog # CHMCAB5510
Supplier:  Chemicon
CAS Number:  
undefined
L(+)-Histidine
Catalog # CHMCAB5510
Supplier:  Chemicon
CAS Number:  

Specifications

  • Pk:
    100 µl
  • Seq:
    1286

Specifications

About this item

 :
 :155.16 g/mol
Merck Index:13,04741
Melting Pt:272…273 °C
Density:1.44 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-745-3
CAS Number:71-00-1
Formula:C₆H₉N₃O₂
MDL Number:MFCD00064315