Order Entry
Switzerland
ContactUsLinkComponent
L(+)-Leucine for synthesis, Sigma-Aldrich®
L(+)-Leucine for synthesis, Sigma-Aldrich®
Catalog # 8.16009.0100
Supplier:  Merck
CAS Number:  
L(+)-Leucine for synthesis, Sigma-Aldrich®
Catalog # 8.16009.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    24 g/l (20 °C)
  • Melting Point:
    300 °C (decomposition)
  • Molar Mass:
    131.18 g/mol
  • Bulk Density:
    300 - 400 kg/m3
  • Density:
    1.29 g/cm3 (20 °C)
  • pH:
    5.5 - 6.5 (20 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :131.17 g/mol
Merck Index:14,05451
Melting Pt:300 °C
Density:1.165 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-522-0
CAS Number:61-90-5
Formula:C₆H₁₃NO₂
MDL Number:MFCD00002617