Order Entry
Switzerland
Orders LinkContactUsLinkComponent
D(-)-Isoleucine

D(-)-Isoleucine

Glass bottle for solids, 100 mg
Catalog # MOLE24366220-100MG
Marketsource
Supplier:   Molekula
D(-)-Isoleucine
Catalog # MOLE24366220-100MG
Supplier:  Molekula
Marketsource
CAS Number:  319-78-8
D(-)-Isoleucine

Glass bottle for solids, 100 mg

CAS Number:  319-78-8
Price
Inquire For Price
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  (2R,3R)-(-)-Isoleucine, H-D-Ile-OH
  • CAS Number:319-78-8
  • Formula:H₃CCH₂CH(CH₃)CH(NH₂)COOH
  • MDL Number:MFCD00064221
  • Molecular Weight:131.17 g/mol
  • Boiling Pt:235 °C (1013 hPa)
  • Melting Pt:98 °C
  • Density:1.224 g/cm³ (20 °C)
  •  :Density: 1.224 g/cm³ (20 °C)
  • EINECS:206-269-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications