Order Entry
Switzerland
ContactUsLinkComponent
(R)-(-)-1-Aminopropan-2-ol
Corrosive
(R)-(-)-1-Aminopropan-2-ol
Catalog # MOLEM48753302
Supplier:  Molekula
CAS Number:  
Corrosive
(R)-(-)-1-Aminopropan-2-ol
Catalog # MOLEM48753302
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :75.11 g/mol
Boiling Pt:160 °C (1013 hPa)
Melting Pt:24…26 °C
Density:0.95 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:220-532-9
UN:3259
ADR:8,II
 :GHS05
CAS Number:2799-16-8
Formula:C₃H₉NO
MDL Number:MFCD00064428