Order Entry
Switzerland
ContactUsLinkComponent
L(+)-Tartaric acid, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Corrosive
L(+)-Tartaric acid, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Corrosive
L(+)-Tartaric acid, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    1390 g/l (20 °C)
  • Melting Point:
    168 - 170 °C
  • Molar Mass:
    150.08 g/mol
  • Bulk Density:
    1000 kg/m3
  • Flash Point:
    210 °C
  • Density:
    1.76 g/cm3 (20 °C)
  • pH:
    1.6 (100 g/l, H2O, 25 °C)
  • Ignition Point:
    425 °C

Specification Test Results

Related Information

Frequently Bought Together